3-(2,5-Dimethylphenylcarbamoyl)-4-fluorobenzeneboronic acid, 97%
H54129.03EA 156 EUR
ALFAH54129.03 ALFAH54129.MD
3-(2,5-Dimethylphenylcarbamoyl)-4-fluorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BFNO3
Formula Weight: 287.09
MDL No.: MFCD20040137