3-Fluoro-4-(4-methylphenylcarbamoyl)benzeneboronic acid, 97%

H54270.03EA 306 EUR
ALFAH54270.03 ALFAH54270.MD
3-Fluoro-4-(4-methylphenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H13BFNO3
Formula Weight: 273.07
MDL No.: MFCD20265220
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR