4-Carboxy-3-fluorobenzeneboronic acid pinacol ester, 98%

H53190.MDEA 15.7 EUR
ALFAH53190.MD ALFAH53190.03
4-Carboxy-3-fluorobenzeneboronic acid pinacol ester, 98%
Purity: 98%
CAS No.: 867256-77-7
Molecular Formula: C13H16BFO4
Formula Weight: 266.08
Melting Point: 289-295°
MDL No.: MFCD09027284
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR