4-Carboxy-2-fluorphenylboronsäurepinakolester, Sigma-Aldrich®
Lieferant: Merck
SIAL720852-1GEA
66.5
EUR
SIAL720852-1G
4-Carboxy-2-fluorphenylboronsäurepinakolester, Sigma-Aldrich®
4-Carboxy-2-fluorphenylboronsäurepinakolester
Formel:
C₁₃H₁₆BFO₄ Molecular Weight: 266,07 g/mol |
MDL:
MFCD12756466 CAS-Nummer: 1050423-87-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...