4-Amino-3-chlorphenol Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL385743-5GEA 52.9 EUR
SIAL385743-5G
4-Amino-3-chlorphenol Hydrochlorid, Sigma-Aldrich®
4-Amino-3-chlorphenol Hydrochlorid
Formel: C₆H₇Cl₂NO
Molecular Weight: 180,04 g/mol
MDL: MFCD00143110
CAS-Nummer: 52671-64-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR