2-Chloro-4-(4-chloro-3-nitrophenylcarbamoyl)benzeneboronic acid, 97%

H54332.03EA 306 EUR
ALFAH54332.03 ALFAH54332.MD
2-Chloro-4-(4-chloro-3-nitrophenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C13H9BCl2N2O5
Formula Weight: 354.94
MDL No.: MFCD27975651
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR