2-Chloro-5-(2,3-dimethylphenylcarbamoyl)benzeneboronic acid, 97%
H54727.03EA 306 EUR
ALFAH54727.03 ALFAH54727.MD
2-Chloro-5-(2,3-dimethylphenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BClNO3
Formula Weight: 303.55
MDL No.: MFCD22125084