3-Chloro-4-(2,5-dimethylphenylcarbamoyl)benzeneboronic acid, 97%

H54659.MDEA 31.9 EUR
ALFAH54659.MD ALFAH54659.03
3-Chloro-4-(2,5-dimethylphenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BClNO3
Formula Weight: 303.55
MDL No.: MFCD20265312
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR