4-Ethoxycarbonyl-2-nitrobenzeneboronic acid, 97%

H52912.MDEA 13.6 EUR
ALFAH52912.MD ALFAH52912.03
4-Ethoxycarbonyl-2-nitrobenzeneboronic acid, 97%
Purity: 97%
CAS No.: 5785-70-6
Molecular Formula: C9H10BNO6
Formula Weight: 238.99
Melting Point: 116-118°
MDL No.: MFCD02179460
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR