(R)-(+)-N-Benzyl-α-methylbenzylamin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL431737-50MLEA 354 EUR
SIAL431737-50ML SIAL431737-10ML
(R)-(+)-N-Benzyl-α-methylbenzylamin, Sigma-Aldrich®
(R)-(+)-N-Benzyl-α-methylbenzylamin
Formel: C₁₅H₁₇N
Molecular Weight: 211,31 g/mol
MDL: MFCD00015010
CAS-Nummer: 38235-77-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR