Dodecyl-methylsulfid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL641480-25GEA 249 EUR
SIAL641480-25G
Dodecyl-methylsulfid, Sigma-Aldrich®
Dodecyl-methylsulfid
Formel: C₁₃H₂₈S
Molecular Weight: 216,41
MDL: MFCD00015073
CAS-Nummer: 3698-89-3
EINECS: 223-028-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR