Methyl-propyldisulfid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL322539-1GEA 12.7 EUR
SIAL322539-1G
Methyl-propyldisulfid, Sigma-Aldrich®
Methyl-propyldisulfid
Formel: C₄H₁₀S₂
MDL: MFCD00013400
CAS-Nummer: 2179-60-4
EINECS: 218-551-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR