N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylendiamin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL122262-1LEA
95.2
EUR
SIAL122262-1L
N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylendiamin, Sigma-Aldrich®
N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylendiamin
Formel:
C₁₄H₃₂N₂O₄ Molecular Weight: 292,42 |
MDL:
MFCD00004534 CAS-Nummer: 102-60-3 EINECS: 203-041-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...