DL-Methioninsulfoxid, Sigma-Aldrich®

Lieferant: Merck

SIAL64430-5G-FEA 67.9 EUR
SIAL64430-5G-F
DL-Methioninsulfoxid, Sigma-Aldrich®
DL-Methioninsulfoxid
Formel: C₅H₁₁NO₃S
Molecular Weight: 165,21 g/mol
MDL: MFCD00002620
CAS-Nummer: 62697-73-8
EINECS: 263-700-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR