Aceton-[2-13C] (99% ¹³C ), Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ quality level = MQ200

SIAL299197-1GEA 924 EUR
SIAL299197-1G SIAL299197-250MG
Aceton-[2-13C] (99% ¹³C ), Sigma-Aldrich®
Aceton-[2-13C]
Formel: C₃H₆O
Lagertemperatur: Raumtemperatur
MDL: MFCD00069967
CAS-Nummer: 3881-06-9
UN: 1090
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR