2-Methyl-5-nitrobenzothiazol ≥98%

Lieferant: Thermo Scientific

B25354.14EA 415 EUR
B25354.14
2-Methyl-5-nitrobenzothiazol ≥98%
2-Methyl-5-nitrobenzothiazole
Formel: C₈H₆N₂O₂S
Schmelzpunkt: 132…134 °C
MDL: MFCD00602735
CAS-Nummer: 2941-66-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR