1-(Phenylsulfonyl)-6-indolboronsäure
Lieferant: COMBI-BLOCKS INC.
COBBFA-1082-1GEA
255
EUR
COBBFA-1082-1G
1-(Phenylsulfonyl)-6-indolboronsäure
1-(Phenylsulphonyl)-6-indoleboronic acid
Formel:
C₁₄H₁₂BNO₄S Molecular Weight: 301,1254 g/mol |
MDL:
MFCD03840611 CAS-Nummer: 480438-52-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...