1-(Phenylsulfonyl)-3-indolboronsäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL563870-1GEA
74.9
EUR
SIAL563870-1G
1-(Phenylsulfonyl)-3-indolboronsäure, Sigma-Aldrich®
1-(Phenylsulphonyl)-3-indoleboronic acid
Formel:
C₁₄H₁₂BNO₄S Molecular Weight: 301,1300 g/mol |
MDL:
MFCD02681892 CAS-Nummer: 129271-98-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...