1-(Phenylsulfonyl)-3-indolboronsäure
Lieferant: Thermo Scientific
MAYBCC03112DAEA
0
EUR
MAYBCC03112DA
MAYBCC03112CB
1-(Phenylsulfonyl)-3-indolboronsäure
1-(Phenylsulphonyl)-3-indoleboronic acid
Formel:
C₁₄H₁₂BNO₄S Molecular Weight: 301,1300 g/mol |
MDL:
MFCD02681892 CAS-Nummer: 129271-98-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...