2-Indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS INC.

COBBFM-2368-1GEA 83.2 EUR
COBBFM-2368-1G
2-Indolboronsäurepinakolester
2-Indolboronsäurepinakolester
Formel: C₁₄H₁₈BNO₂
MDL: MFCD09751284
CAS-Nummer: 476004-81-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR