4'-(1-Imidazolyl)acetophenon, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL183733-5GEA
92.2
EUR
SIAL183733-5G
4'-(1-Imidazolyl)acetophenon, Sigma-Aldrich®
4'-(1-Imidazolyl)acetophenon
Formel:
C₁₁H₁₀N₂O Molecular Weight: 186,21 g/mol |
MDL:
MFCD00005282 CAS-Nummer: 10041-06-2 EINECS: 233-123-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...