4'-(1-Imidazolyl)acetophenon

Lieferant: CHEMPUR

CHMPFL13609.POREA 0 EUR
CHMPFL13609.POR
4'-(1-Imidazolyl)acetophenon
4'-(1-Imidazolyl)acetophenon
Formel: C₁₁H₁₀N₂O
Molecular Weight: 186,21 g/mol
MDL: MFCD00005282
CAS-Nummer: 10041-06-2
EINECS: 233-123-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR