2-Amino-3-formyl-6-methylchromon

Lieferant: CHEMPUR

FLUO012026.1GEA 113 EUR
FLUO012026.1G CHMPFL12026.POR
2-Amino-3-formyl-6-methylchromon
2-Amino-3-formyl-6-methylchromon
Formel: C₁₁H₉NO₃
MDL: MFCD00138945
CAS-Nummer: 68301-75-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR