2-Amino-3-formyl-6-isopropylchromon

Lieferant: CHEMPUR

CHMPFL12030.POREA 0 EUR
CHMPFL12030.POR
2-Amino-3-formyl-6-isopropylchromon
2-Amino-3-formyl-6-isopropylchromon
Formel: C₁₃H₁₃NO₃
MDL: MFCD07368730
CAS-Nummer: 887266-85-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR