Methylchromon-2-carboxylat

Lieferant: CHEMPUR

CHMPFL12064.POREA 0 EUR
CHMPFL12064.POR
Methylchromon-2-carboxylat
Methylchromon-2-carboxylat
Formel: C₁₁H₈O₄
MDL: MFCD03424511
CAS-Nummer: 27455-31-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR