Methyl-6-bromchromon-2-carboxylat

Lieferant: CHEMPUR

CHMPFL12067.POREA 0 EUR
CHMPFL12067.POR
Methyl-6-bromchromon-2-carboxylat
Methyl-6-bromchromon-2-carboxylat
Formel: C₁₁H₇BrO₄
Molecular Weight: 283,077 g/mol
MDL: MFCD03424512
CAS-Nummer: 116543-92-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR