Tetramethytetraselenafulvalen, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL274402-50MGEA 87.4 EUR
SIAL274402-50MG
Tetramethytetraselenafulvalen, Sigma-Aldrich®
Tetramethytetraselenafulvalen
Formel: C₁₀H₁₂Se₂
MDL: MFCD00011507
CAS-Nummer: 55259-49-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR