trans-4-(Aminomethyl)cyclohexancarbonsäure

Lieferant: U.S. Pharmacopeia

USPH1672745EA 353 EUR
USPH1672745
trans-4-(Aminomethyl)cyclohexancarbonsäure
trans-4-(Aminomethyl)cyclohexanecarboxylic acid
Formel: C₈H₁₅NO₂
Molecular Weight: 157,21 g/mol
MDL: MFCD00001466
CAS-Nummer: 1197-18-8
EINECS: 214-818-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR