2-Butoxy-5-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-6365-5GEA 135 EUR
COBBBB-6365-5G
2-Butoxy-5-(trifluormethyl)phenylboronsäure
2-Butoxy-5-(trifluormethyl)phenylboronsäure
Formel: C₁₁H₁₄BF₃O₃
MDL: MFCD16036115
CAS-Nummer: 1256345-98-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR