3-Chlor-5-methylphenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-3598-1GEA 117 EUR
COBBBB-3598-1G COBBBB-3598-5G
3-Chlor-5-methylphenylboronsäure
3-Chlor-5-methylphenylboronsäure
Formel: C₇H₈BClO₂
Molecular Weight: 170,40 g/mol
MDL: MFCD09027241
CAS-Nummer: 913836-14-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR