3-Chlor-5-methylphenylboronsäure ≥98%

Lieferant: Thermo Scientific
Achtung

H53179.03EA 253 EUR
H53179.03
3-Chlor-5-methylphenylboronsäure ≥98%
3-Chlor-5-methylphenylboronsäure
Formel: C₇H₈BClO₂
Molecular Weight: 170,40
Schmelzpunkt: 235…238 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027241
CAS-Nummer: 913836-14-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR