4-Isobutyl-3-nitrobenzeneboronic acid, 97%

H53152.MDEA 99.3 EUR
ALFAH53152.MD ALFAH53152.03
4-Isobutyl-3-nitrobenzeneboronic acid, 97%
Purity: 97%
CAS No.: 850568-57-9
Molecular Formula: C10H14BNO4
Formula Weight: 223.04
Melting Point: 168-174°
MDL No.: MFCD04115653
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR