1-Decylboronsäurepinakolester ≥98%

Lieferant: Alfa Aesar

H30403.14EA 490 EUR
H30403.14 H30403.03 H30403.06
1-Decylboronsäurepinakolester ≥98%
1-Decylboronsäurepinakolester
Formel: C₁₆H₃₃BO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12546186
CAS-Nummer: 141091-38-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR