6-Brom-2-methylchromon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL550329-1GEA 96.2 EUR
SIAL550329-1G
6-Brom-2-methylchromon, Sigma-Aldrich®
6-Brom-2-methylchromon
Formel: C₁₀H₇BrO₂
MDL: MFCD03701517
CAS-Nummer: 30779-63-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR