6-Fluor-o-toluidin

Lieferant: Apollo Scientific

APOSPC1391-5GEA 17.6 EUR
APOSPC1391-5G
6-Fluor-o-toluidin
6-Fluor-o-toluidin
Formel: C₇H₈FN
MDL: MFCD06658252
CAS-Nummer: 443-89-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR