2-Amino-4-(3-pyridyl)thiazol 97%
Lieferant: Thermo Scientific
|
Achtung
|
H58630.04EA
304
EUR
H58630.04
2-Amino-4-(3-pyridyl)thiazol 97%
2-Amino-4-(3-pyridyl)thiazole
Formel:
C₈H₇N₃S Molecular Weight: 177,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00218013 CAS-Nummer: 30235-27-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...