ddNTP, 100 mM solutions
Lieferant: Cytiva
27-2081-01EA 301 EUR
27-2081-01 27-2061-01
ddNTP, 100 mM solutions
Nukleinsäure-Reagenzien Nukleotide
Easy to use, time saving solutions for use in any application requiring ddNTPs of high purity, such as dideoxy sequencing and genotyping reactions.
  • High purity ≥98% triphosphate
  • Each lot of ddNTPs is functionally tested to meet or exceed criteria for high quality sequencing