Acrylamide solutions, Fluorescent NEXT GEL®
M290-500ML-KITEA 0 EUR
VWRCM290-500ML-KIT VWRCM291-100ML-KIT
Acrylamide solutions, Fluorescent NEXT GEL®
Pre-mixed, ready to pour acrylamide solution for the SDS- PAGE fluorescent analysis of proteins.
  • Ready-to-pour SDS polyacrylamide solutions - just add APS and TEMED
  • Faster gel casting with no stacking gel required
  • Convenient fluorescent dye stains proteins during electrophoresis and requires no destaining post-electrophoresis
  • Broad range of separation
  • High resolution of protein bands
Lieferumfang: Kit includes Fluorescent NEXT GEL® 10% solution and NEXT GEL® running buffer, 20X.