4-Mercapto-3-nitrobenzotrifluorid

Lieferant: CHEMPUR

CHMPFL13864.POREA 0 EUR
CHMPFL13864.POR
4-Mercapto-3-nitrobenzotrifluorid
4-Mercapto-3-nitrobenzotrifluorid
Formel: C₇H₄F₃NO₂S
MDL: MFCD00013454
CAS-Nummer: 14371-82-5
EINECS: 238-343-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR