2'-Isopropylacetanilid

Lieferant: Thermo Scientific

MAYBTL00848.50EA 0 EUR
MAYBTL00848.50
2'-Isopropylacetanilid
2'-Isopropylacetanilid
Formel: C₁₁H₁₅NO
MDL: MFCD00833396
CAS-Nummer: 19246-04-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR