(4-Nitrophenyl)iodacetat, Sigma-Aldrich®

Lieferant: Merck

SIAL57865-5G-FEA 62.7 EUR
SIAL57865-5G-F
(4-Nitrophenyl)iodacetat, Sigma-Aldrich®
(4-Nitrophenyl)iodacetat
Formel: C₈H₆INO₄
CAS-Nummer: 31252-85-4
EINECS: 250-536-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR