(3R)-Tetrahydro-3-furanol

Lieferant: COMBI-BLOCKS INC.

COBBSS-6020-5GEA 92.4 EUR
COBBSS-6020-5G
(3R)-Tetrahydro-3-furanol
(R)-(-)-3-Hydroxytetrahydrofuran
Formel: C₄H₈O₂
Molecular Weight: 88,11 g/mol
MDL: MFCD00067101
CAS-Nummer: 86087-24-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR