α,α,α,2,6-Pentafluortoluol

Lieferant: COMBI-BLOCKS INC.

COBBOR-5680-25GEA 431 EUR
COBBOR-5680-25G
α,α,α,2,6-Pentafluortoluol
α,α,α,2,6-Pentafluortoluol
Formel: C₇H₃F₅
MDL: MFCD03412213
CAS-Nummer: 64248-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR