Ethinyltributylzinn

Lieferant: FLUOROCHEM LTD (CHEMPUR)
Gefahr

FLUOSNE4900.10GEA 620 EUR
FLUOSNE4900.10G
Ethinyltributylzinn
Ethynyltributyltin
Formel: C₁₄H₂₈Sn
Molecular Weight: 315,09 g/mol
MDL: MFCD00009420
CAS-Nummer: 994-89-8
UN: 2788
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR