5-Amino-1-methyl-1H-pyrazol

Lieferant: CHEMPUR

CHMPCFF791.EA 0 EUR
FLUO046179.5G CHMPCFF791. FLUO030129.1G FLUO030129.5G
5-Amino-1-methyl-1H-pyrazol
5-Amino-1-methyl-1H-pyrazole
Formel: C₄H₇N₃
Molecular Weight: 97,12 g/mol
MDL: MFCD00068156
CAS-Nummer: 1192-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR