Hafnium(IV)-tert-butanolat ≥99,9% (Hf-Basis) <1.5%-Zr

Lieferant: ABCR GMBH
Achtung

ABCRAB106072.0010EA 0 EUR
ABCRAB106072.0010
Hafnium(IV)-tert-butanolat ≥99,9% (Hf-Basis) <1.5%-Zr
Hafnium(IV)-tert-butanolat

Alkoxide, Metal Alkoxides, Volatile Precursors for CVD

Formel: C₁₆H₃₆HfO₄
Molecular Weight: 470,94 g/mol
Siedepunkt: 90 °C (5 mmHg)
Schmelzpunkt: 8 °C
Dichte: 1,166
MDL: MFCD00070458
CAS-Nummer: 2172-02-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR