(±)-Ethyl-2-chlorpropionat

Lieferant: EHRENSTORFER GMBH

EHERC11502800EA 48 EUR
EHERC11502800
(±)-Ethyl-2-chlorpropionat
(±)-Ethyl-2-chlorpropionat
Formel: C₅H₉ClO₂
Molecular Weight: 136,58 g/mol
MDL: MFCD00000864
CAS-Nummer: 535-13-7
EINECS: 208-610-0
UN: 2935
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR