3-Brom-5-fluorbenzaldehyd
Lieferant: COMBI-BLOCKS INC.
COBBOT-1261-100GEA
826
EUR
COBBOT-1261-100G
COBBOT-1261-25G
3-Brom-5-fluorbenzaldehyd
3-Bromo-5-fluorobenzaldehyde
Formel:
C₇H₄BrFO |
MDL:
MFCD04116319 CAS-Nummer: 188813-02-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...