2-Thiophenboronsäure ≥97%

Lieferant: ABCR GMBH
Achtung

Synonyme: 2-ThienylboronsäureThiophen-2-boronsäure

ABCRAB130592.0005EA 0 EUR
ABCRAB130592.0005
2-Thiophenboronsäure ≥97%
2-Thiopheneboronic acid

Boronic Acid and Ester

Formel: C₄H₅BO₂S
Molecular Weight: 127,96 g/mol
Schmelzpunkt: 138…140 °C
MDL: MFCD00151850
CAS-Nummer: 6165-68-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR