2-Bromcumol

Lieferant: COMBI-BLOCKS INC.

COBBOT-0471-100GEA 318 EUR
COBBOT-0471-100G
2-Bromcumol
2-Bromcumol
Formel: C₉H₁₁Br
Molecular Weight: 199,09 g/mol
MDL: MFCD00051567
CAS-Nummer: 7073-94-1
EINECS: 230-370-0
UN: 3082
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR